siRNA Details
| Gene Symbol: Mki67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271366.1 | NP_001258295.1 | 13 | 6560 | ||
| XM_006230453.2 | XP_006230515.1 | 12 | 6909 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mki67| Gene Symbol: Mki67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271366.1 | NP_001258295.1 | 13 | 6560 | ||
| XM_006230453.2 | XP_006230515.1 | 12 | 6909 | |||