siRNA Details
| Gene Symbol: Actn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170325.1 | NP_001163796.1 | 9 | 990 | ||
| GenBank mRNA | DY315226.1 | - | 5 | 320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actn2| Gene Symbol: Actn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170325.1 | NP_001163796.1 | 9 | 990 | ||
| GenBank mRNA | DY315226.1 | - | 5 | 320 | ||