siRNA Details
| Gene Symbol: Usp6nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106120.1 | NP_001099590.1 | 3 | 268 | ||
| XM_006254232.2 | XP_006254294.1 | 2 | 512 | |||
| XM_006254233.1 | XP_006254295.1 | 2 | 137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp6nl| Gene Symbol: Usp6nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106120.1 | NP_001099590.1 | 3 | 268 | ||
| XM_006254232.2 | XP_006254294.1 | 2 | 512 | |||
| XM_006254233.1 | XP_006254295.1 | 2 | 137 | |||