siRNA Details
| Gene Symbol: Nmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207590.1 | NP_997473.1 | 1 | 76 | ||
| XM_008771854.1 | XP_008770076.1 | 1 | 94 | |||
| XM_008771855.1 | XP_008770077.1 | 1 | 191 | |||
| XM_008771856.1 | XP_008770078.1 | 1 | 94 | |||
| GenBank mRNA | AJ629217.1 | - | 1 | 76 | ||

Nmt2
Genomic Map