siRNA Details
| Gene Symbol: Trdmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031643.1 | NP_001026813.1 | 2 | 150 | ||
| GenBank mRNA | BC098700.1 | - | 2 | 150 | 7387075 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trdmt1| Gene Symbol: Trdmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031643.1 | NP_001026813.1 | 2 | 150 | ||
| GenBank mRNA | BC098700.1 | - | 2 | 150 | 7387075 | |