siRNA Details
| Gene Symbol: St8sia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213624.1 | NP_998789.1 | 208 | |||
| GenBank mRNA | AJ699423.1 | - | 208 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St8sia6| Gene Symbol: St8sia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213624.1 | NP_998789.1 | 208 | |||
| GenBank mRNA | AJ699423.1 | - | 208 | |||