siRNA Details
Gene Symbol: Mrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106123.2 | NP_001099593.2 | 25 | 3679 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106123.2 | NP_001099593.2 | 25 | 3679 |