siRNA Details
| Gene Symbol: Slc39a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106124.1 | NP_001099594.1 | 2 | 436 | ||
| XM_006254285.2 | XP_006254347.1 | 4 | 1072 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc39a12| Gene Symbol: Slc39a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106124.1 | NP_001099594.1 | 2 | 436 | ||
| XM_006254285.2 | XP_006254347.1 | 4 | 1072 | |||