siRNA Details
| Gene Symbol: Commd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198732.2 | NP_942027.1 | 5 | 441 | ||
| GenBank mRNA | BC060591.1 | - | 5 | 444 | 6920278 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Commd3| Gene Symbol: Commd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198732.2 | NP_942027.1 | 5 | 441 | ||
| GenBank mRNA | BC060591.1 | - | 5 | 444 | 6920278 | |