siRNA Details
| Gene Symbol: Cndp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010920.1 | NP_001010920.1 | 9 | 1106 | ||
| XM_006254998.2 | XP_006255060.1 | 8 | 1152 | |||
| XM_006254999.2 | XP_006255061.1 | 9 | 1146 | |||
| GenBank mRNA | AY569013.1 | - | 9 | 1106 | ||
| BC095904.1 | - | 8 | 1079 | 7110846 | ||

Cndp2
Genomic Map