siRNA Details
| Gene Symbol: Zfp236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255011.2 | XP_006255073.1 | 11 | 1838 | ||
| XM_006255013.2 | XP_006255075.1 | 11 | 1838 | |||
| XM_006255016.2 | XP_006255078.1 | 11 | 1838 | |||
| XM_008772218.1 | XP_008770440.1 | 11 | 1972 | |||
| XM_008772219.1 | XP_008770441.1 | 11 | 1838 | |||
| XM_008772220.1 | XP_008770442.1 | 11 | 1838 | |||

Zfp236
Genomic Map