siRNA Details
| Gene Symbol: Zfp236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255011.2 | XP_006255073.1 | 27 | 4928 | ||
| XM_006255013.2 | XP_006255075.1 | 26 | 4748 | |||
| XM_008772218.1 | XP_008770440.1 | 27 | 5062 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp236| Gene Symbol: Zfp236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255011.2 | XP_006255073.1 | 27 | 4928 | ||
| XM_006255013.2 | XP_006255075.1 | 26 | 4748 | |||
| XM_008772218.1 | XP_008770440.1 | 27 | 5062 | |||