siRNA Details
| Gene Symbol: Ctdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106131.1 | NP_001099601.1 | 4 | 674 | ||
| XM_006254942.2 | XP_006255004.1 | 4 | 1779 | |||
| XM_006254943.2 | XP_006255005.1 | 4 | 1780 | |||
| XM_006254944.2 | XP_006255006.1 | 4 | 1781 | |||

Ctdp1
Genomic Map