siRNA Details
| Gene Symbol: Rpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201415.1 | NP_958818.1 | 4 | 220 | ||
| GenBank mRNA | X60212.1 | - | 4 | 220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl17| Gene Symbol: Rpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201415.1 | NP_958818.1 | 4 | 220 | ||
| GenBank mRNA | X60212.1 | - | 4 | 220 | ||