siRNA Details
| Gene Symbol: Lipg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012741.1 | NP_001012759.1 | 2 | 133 | ||
| GenBank mRNA | AY916123.1 | - | 2 | 133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lipg| Gene Symbol: Lipg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012741.1 | NP_001012759.1 | 2 | 133 | ||
| GenBank mRNA | AY916123.1 | - | 2 | 133 | ||