siRNA Details
| Gene Symbol: Prrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033887.1 | NP_001029059.1 | 5, 6 | 900 | ||
| XM_006254746.2 | XP_006254808.1 | 6, 7 | 1096 | |||
| GenBank mRNA | BC101903.1 | - | 5, 6 | 900 | 7934311 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prrc1| Gene Symbol: Prrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033887.1 | NP_001029059.1 | 5, 6 | 900 | ||
| XM_006254746.2 | XP_006254808.1 | 6, 7 | 1096 | |||
| GenBank mRNA | BC101903.1 | - | 5, 6 | 900 | 7934311 | |