siRNA Details
Gene Symbol: Poli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106137.1 | NP_001099607.1 | 2 | 207 | ||
XM_006254900.2 | XP_006254962.1 | 2 | 197 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Poli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106137.1 | NP_001099607.1 | 2 | 207 | ||
XM_006254900.2 | XP_006254962.1 | 2 | 197 |