siRNA Details
| Gene Symbol: Poli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106137.1 | NP_001099607.1 | 3 | 288 | ||
| XM_006254900.2 | XP_006254962.1 | 3 | 278 | |||
| XM_006254901.2 | XP_006254963.1 | 2 | 151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Poli| Gene Symbol: Poli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106137.1 | NP_001099607.1 | 3 | 288 | ||
| XM_006254900.2 | XP_006254962.1 | 3 | 278 | |||
| XM_006254901.2 | XP_006254963.1 | 2 | 151 | |||