siRNA Details
| Gene Symbol: Psmg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106138.2 | NP_001099608.2 | 4, 5 | 482 | ||
| XM_008772117.1 | XP_008770339.1 | 4, 5 | 511 | |||
| GenBank mRNA | DV720222.1 | - | 3, 4 | 212 | ||
| FQ212569.1 | - | 4, 5 | 546 | |||
| FQ230678.1 | - | 4, 5 | 510 | |||

Psmg2
Genomic Map