siRNA Details
| Gene Symbol: Nedd4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008300.2 | NP_001008301.2 | 22 | 2156 | ||
| GenBank mRNA | BC086371.1 | - | 6 | 678 | ||
| GQ160815.1 | - | 21 | 2096 | |||
| GQ160816.1 | - | 21 | 2096 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nedd4l| Gene Symbol: Nedd4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008300.2 | NP_001008301.2 | 22 | 2156 | ||
| GenBank mRNA | BC086371.1 | - | 6 | 678 | ||
| GQ160815.1 | - | 21 | 2096 | |||
| GQ160816.1 | - | 21 | 2096 | |||