siRNA Details
| Gene Symbol: Nars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025635.2 | NP_001020806.2 | 14, 15 | 1602 | ||
| NM_001142949.1 | NP_001136421.1 | 12, 13 | 1449 | |||
| XM_006254844.2 | XP_006254906.1 | 14, 15 | 1660 | |||
| GenBank mRNA | AY387099.1 | - | 11, 12 | 1355 | ||
| BC099099.1 | - | 13, 14 | 1483 | 7459693 | ||

Nars
Genomic Map