siRNA Details
Gene Symbol: Ppargc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176075.2 | NP_788264.1 | 1145 | |||
GenBank mRNA | AY188951.1 | - | 1145 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ppargc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176075.2 | NP_788264.1 | 1145 | |||
GenBank mRNA | AY188951.1 | - | 1145 |