siRNA Details
Gene Symbol: Tcof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106143.1 | NP_001099613.1 | 20 | 3267 | ||
XM_008772124.1 | XP_008770346.1 | 20 | 3250 | |||
GenBank mRNA | FQ232316.1 | - | 2 | 290 |
