siRNA Details
| Gene Symbol: Chsy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_225912.7 | XP_225912.2 | 2872 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001081328, AK019528, BC157985, BC151164, BC151165, and others.

Chsy3
Genomic Map