siRNA Details
| Gene Symbol: Slc27a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106145.1 | NP_001099615.1 | 5 | 1265 | ||
| Gene Symbol: LOC100910486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751788.3 | XP_003751836.2 | 1 | 1658 | ||
| Gene Symbol: LOC103694868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773817.1 | XP_008772039.1 | 1 | 1658 | ||

Slc27a6
Genomic Map