siRNA Details
| Gene Symbol: Trim36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106147.1 | NP_001099617.1 | 8 | 1380 | ||
| XM_006254686.2 | XP_006254748.1 | 8 | 1608 | |||
| XM_008772127.1 | XP_008770349.1 | 8 | 1608 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim36| Gene Symbol: Trim36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106147.1 | NP_001099617.1 | 8 | 1380 | ||
| XM_006254686.2 | XP_006254748.1 | 8 | 1608 | |||
| XM_008772127.1 | XP_008770349.1 | 8 | 1608 | |||