siRNA Details
| Gene Symbol: Lars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009637.1 | NP_001009637.1 | 9 | 914 | ||
| XM_006254672.2 | XP_006254734.1 | 9 | 1020 | |||
| GenBank mRNA | BC087655.1 | - | 9 | 914 | 7312531 | |
| FQ212494.1 | - | 9 | 951 | |||

Lars
Genomic Map