siRNA Details
| Gene Symbol: Pcdhga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037137.1 | NP_001032214.1 | 1 | 917 | ||
| GenBank mRNA | AY574016.1 | - | 1 | 917 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhga5| Gene Symbol: Pcdhga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037137.1 | NP_001032214.1 | 1 | 917 | ||
| GenBank mRNA | AY574016.1 | - | 1 | 917 | ||