siRNA Details
| Gene Symbol: Cxxc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007628.2 | NP_001007629.1 | 1 | 367 | ||
| GenBank mRNA | BC083622.1 | - | 1 | 367 | 7134100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxxc5| Gene Symbol: Cxxc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007628.2 | NP_001007629.1 | 1 | 367 | ||
| GenBank mRNA | BC083622.1 | - | 1 | 367 | 7134100 | |