siRNA Details
| Gene Symbol: Mzb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024240.1 | NP_001019411.1 | 3, 4 | 419 | ||
| GenBank mRNA | BC093397.1 | - | 3, 4 | 419 | 7389084 | |
| FQ234743.1 | - | 3, 4 | 453 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mzb1| Gene Symbol: Mzb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024240.1 | NP_001019411.1 | 3, 4 | 419 | ||
| GenBank mRNA | BC093397.1 | - | 3, 4 | 419 | 7389084 | |
| FQ234743.1 | - | 3, 4 | 453 | |||