siRNA Details
| Gene Symbol: Dnajc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013887.1 | NP_001013909.1 | 9 | 1075 | ||
| XM_006254546.2 | XP_006254608.1 | 8 | 1054 | |||
| GenBank mRNA | BC079289.1 | - | 9 | 1075 | 7134364 | |
| DQ158861.1 | - | 8 | 1008 | |||
| FQ214241.1 | - | 8 | 928 | |||

Dnajc18
Genomic Map