siRNA Details
| Gene Symbol: Wnt8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106155.1 | NP_001099625.1 | 1, 2 | 125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt8a| Gene Symbol: Wnt8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106155.1 | NP_001099625.1 | 1, 2 | 125 | ||