siRNA Details
Gene Symbol: Brd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008509.1 | NP_001008509.1 | 17 | 2522 | ||
XM_006254562.2 | XP_006254624.1 | 18 | 2708 | |||
XM_006254563.2 | XP_006254625.1 | 18 | 2709 | |||
XM_006254564.2 | XP_006254626.1 | 17 | 2485 | |||
XM_008772029.1 | XP_008770251.1 | 18 | 2746 | |||
GenBank mRNA | AB180485.1 | - | 17 | 2522 |
