siRNA Details
| Gene Symbol: Brd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008509.1 | NP_001008509.1 | 11 | 1402 | ||
| XM_006254562.2 | XP_006254624.1 | 12 | 1588 | |||
| XM_006254563.2 | XP_006254625.1 | 12 | 1589 | |||
| XM_006254564.2 | XP_006254626.1 | 11 | 1365 | |||
| XM_008772029.1 | XP_008770251.1 | 12 | 1626 | |||
| GenBank mRNA | AB180485.1 | - | 11 | 1402 | ||

Brd8
Genomic Map