siRNA Details
| Gene Symbol: Fam13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106158.1 | NP_001099628.1 | 12 | 1857 | ||
| XM_008772030.1 | XP_008770252.1 | 12 | 1547 | |||
| XM_008772031.1 | XP_008770253.1 | 12 | 1547 | |||
| XM_008772032.1 | XP_008770254.1 | 12 | 1547 | |||
| XM_008772033.1 | XP_008770255.1 | 11 | 1481 | |||
| GenBank mRNA | FQ228342.1 | - | 10 | 1369 | ||

Fam13b
Genomic Map