siRNA Details
| Gene Symbol: Rprd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772003.1 | XP_008770225.1 | 5 | 647 | ||
| XM_214609.8 | XP_214609.6 | 5 | 647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rprd1a| Gene Symbol: Rprd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772003.1 | XP_008770225.1 | 5 | 647 | ||
| XM_214609.8 | XP_214609.6 | 5 | 647 | |||