siRNA Details
| Gene Symbol: Ino80c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017446.1 | NP_001017446.1 | 1 | 154 | ||
| GenBank mRNA | BC079333.1 | - | 1 | 62 | ||
| BC091281.1 | - | 1 | 154 | 7192011 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ino80c| Gene Symbol: Ino80c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017446.1 | NP_001017446.1 | 1 | 154 | ||
| GenBank mRNA | BC079333.1 | - | 1 | 62 | ||
| BC091281.1 | - | 1 | 154 | 7192011 | ||