siRNA Details
Gene Symbol: LOC103689930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254461.2 | XP_006254523.1 | 11 | 1846 | ||
XM_006254462.2 | XP_006254524.1 | 11 | 1843 | |||
XM_008771994.1 | XP_008770216.1 | 11 | 1900 |
Gene Symbol: Trappc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106160.1 | NP_001099630.1 | 1600 |

More Information
Other Species Targeted

Gene Ontology Categories:
