siRNA Details
| Gene Symbol: LOC103689930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254461.2 | XP_006254523.1 | 11, 12 | 1871 | ||
| XM_006254462.2 | XP_006254524.1 | 11, 12 | 1868 | |||
| XM_008771994.1 | XP_008770216.1 | 11, 12 | 1925 | |||
| Gene Symbol: Trappc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106160.1 | NP_001099630.1 | 1625 | |||

LOC103689930
Genomic Map