siRNA Details
| Gene Symbol: Dsg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191079.1 | NP_001178008.1 | 8 | 946 | ||
| XM_008771934.1 | XP_008770156.1 | 8 | 946 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dsg3| Gene Symbol: Dsg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191079.1 | NP_001178008.1 | 8 | 946 | ||
| XM_008771934.1 | XP_008770156.1 | 8 | 946 | |||