siRNA Details
| Gene Symbol: Dsg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199490.1 | NP_955784.1 | 11 | 1543 | ||
| GenBank mRNA | AY314982.1 | - | 11 | 1543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dsg4| Gene Symbol: Dsg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199490.1 | NP_955784.1 | 11 | 1543 | ||
| GenBank mRNA | AY314982.1 | - | 11 | 1543 | ||