siRNA Details
| Gene Symbol: Chst9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170470.1 | NP_001163941.1 | 4 | 761 | ||
| XM_006254435.2 | XP_006254497.1 | 3 | 1390 | |||
| XM_008771936.1 | XP_008770158.1 | 2 | 1295 | |||
| XM_008771937.1 | XP_008770159.1 | 3 | 5769 | |||

Chst9
Genomic Map