siRNA Details
| Gene Symbol: Snrpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106163.1 | NP_001099633.1 | 2, 3 | 205 | ||
| GenBank mRNA | BC167095.1 | - | 2, 3 | 98 | 9091450 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snrpd1| Gene Symbol: Snrpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106163.1 | NP_001099633.1 | 2, 3 | 205 | ||
| GenBank mRNA | BC167095.1 | - | 2, 3 | 98 | 9091450 | |