siRNA Details
| Gene Symbol: Usp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008301.1 | NP_001008302.1 | 5 | 461 | ||
| XM_006254396.2 | XP_006254458.1 | 5 | 655 | |||
| XM_006254398.2 | XP_006254460.1 | 4 | 449 | |||
| GenBank mRNA | BC085947.1 | - | 5 | 461 | 7130090 | |

Usp14
Genomic Map