siRNA Details
| Gene Symbol: Thoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047850.1 | NP_001041315.1 | 12 | 1182 | ||
| XM_006254399.2 | XP_006254461.1 | 20 | 1891 | |||
| GenBank mRNA | AY387056.1 | - | 12 | 1182 | ||
| FQ229414.1 | - | 10 | 837 | |||
| Gene Symbol: Da2-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325254.1 | - | 4 | 730 | ||

Thoc1
Genomic Map