siRNA Details
| Gene Symbol: Setd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106167.1 | NP_001099637.1 | 6 | 1054 | ||
| XM_006255070.2 | XP_006255132.1 | 7 | 2004 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Setd6| Gene Symbol: Setd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106167.1 | NP_001099637.1 | 6 | 1054 | ||
| XM_006255070.2 | XP_006255132.1 | 7 | 2004 | |||