siRNA Details
| Gene Symbol: Prss54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013888.1 | NP_001013910.1 | 7 | 1364 | ||
| XM_006255073.2 | XP_006255135.1 | 7 | 1353 | |||
| XM_008772273.1 | XP_008770495.1 | 7 | 1251 | |||
| GenBank mRNA | BC079217.1 | - | 7 | 1364 | 7131896 | |

Prss54
Genomic Map