siRNA Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106169.1 | NP_001099639.1 | 6 | 852 | ||
| XM_006255079.2 | XP_006255141.1 | 6 | 878 | |||
| XM_006255081.2 | XP_006255143.1 | 6 | 842 | |||
| XM_008772281.1 | XP_008770503.1 | 6 | 1247 | |||

Drc7
Genomic Map