siRNA Details
| Gene Symbol: Rspry1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100945.1 | NP_001094415.1 | 3 | 824 | ||
| GenBank mRNA | AY724539.1 | - | 2 | 467 | ||
| BC129112.1 | - | 3 | 824 | 7112017 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rspry1| Gene Symbol: Rspry1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100945.1 | NP_001094415.1 | 3 | 824 | ||
| GenBank mRNA | AY724539.1 | - | 2 | 467 | ||
| BC129112.1 | - | 3 | 824 | 7112017 | ||