siRNA Details
| Gene Symbol: Nup93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011925.1 | NP_001011925.1 | 15, 16 | 1811 | ||
| XM_008772282.1 | XP_008770504.1 | 13, 14 | 3907 | |||
| GenBank mRNA | BC081925.1 | - | 15, 16 | 1811 | 7113037 | |

Nup93
Genomic Map